Order Entry
Ireland
ContactUsLinkComponent
Anti-ProCollagen Type I Propeptide Mouse Monoclonal Antibody [clone: 6F177]
Anti-ProCollagen Type I Propeptide Mouse Monoclonal Antibody [clone: 6F177]
Catalog # USBIP9005-74E
Supplier:  US Biological
Anti-ProCollagen Type I Propeptide Mouse Monoclonal Antibody [clone: 6F177]
Catalog # USBIP9005-74E
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ProCollagen Type I Propeptide
  • Clonality:
    Monoclonal
  • Clone:
    6F177
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 15mM sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A/G chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: ProCollagen Type I Propeptide
Clonality: monoclonal
Clone: 6F177
Conjugation: unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human