Order Entry
Ireland
ContactUsLinkComponent
Anti-Pregnane X Receptor Rabbit Polyclonal Antibody (Biotin)
Anti-Pregnane X Receptor Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIP6001-30D-BIOT
Supplier:  US Biological
undefined
Anti-Pregnane X Receptor Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIP6001-30D-BIOT
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Pregnane X Receptor
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: Pregnane X Receptor
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human