Order Entry
Ireland
ContactUsLinkComponent
Anti-Phospholipase C, gamma 2 Rabbit Polyclonal Antibody
Anti-Phospholipase C, gamma 2 Rabbit Polyclonal Antibody
  USBIP4074-45H
Anti-Phospholipase C, gamma 2 Rabbit Polyclonal Antibody
  USBIP4074-45H

 

  • Antibody type:
    Primary
  • Antigen name:
    Phospholipase C, gamma 2
  • Antigen symbol:
    PLCG2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    liquid
  • Modification:
    phospho-Tyr1217
  • Storage buffer:
    PBS(without Mg2+ and Ca2+), pH 7,4, 150mMsodium chloride, 0,02% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: Phospholipase C, gamma 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human