Order Entry
Ireland
ContactUsLinkComponent
Anti-Peroxiredoxin, 2-Cys Mouse Monoclonal Antibody [clone: 10K305]
Anti-Peroxiredoxin, 2-Cys Mouse Monoclonal Antibody [clone: 10K305]
Catalog # USBIP3352-02
Supplier:  US Biological
Anti-Peroxiredoxin, 2-Cys Mouse Monoclonal Antibody [clone: 10K305]
Catalog # USBIP3352-02
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Peroxiredoxin, 2-Cys
  • Clonality:
    Monoclonal
  • Clone:
    10K305
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Format:
    ascites
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Peroxiredoxin, 2-Cys
Clonality: monoclonal
Clone: 10K305
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse