Anti-PTH Mouse Monoclonal Antibody [clone: L7-13A-B7]
Catalog # USBIP3108-87A-10
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:parathyroid hormone
- Antigen symbol:PTH
- Clonality:Monoclonal
- Clone:L7-13A-B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:1 to 10
- Storage buffer:PBS, pH 7,4.
- Storage temperature:Cool
- Concentration:~1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:10 µg
- Pk:10 µG
Specifications
About this item
Recommended dilution: ELISA: 50ng/ml; ILMA: 50ng/ml
Type: Primary
Antigen: PTH (parathyroid hormone)
Clonality: monoclonal
Clone: L7-13A-B7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human