Anti-Oncofetal Antigen 5T4 Sheep Polyclonal Antibody (Biotin)
USBIO6450-01B
- Antibody type:Primary
- Antigen name:Oncofetal Antigen 5T4
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Form:lyophilized powder
- Amino acid number:31 to 355
- Storage buffer:PBS,BSA
- Storage temperature:Cool
- Concentration:~0,05 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: Oncofetal Antigen 5T4
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Sheep
Isotype: IgG
Reactivity: Human