Anti-Nuclear Receptor Subfamily 2 Group E Member 1 Mouse Monoclonal Antibody [clone: 9G408]
Catalog # USBIN6889-94E2
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Nuclear receptor subfamily 2, group E, member 1
- Antigen symbol:NR2E1
- Clonality:Monoclonal
- Clone:9G408
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Nuclear Receptor Subfamily 2 Group E Member 1
Clonality: monoclonal
Clone: 9G408
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat