Anti-Nuclear Protein Skip Rabbit Polyclonal Antibody
Catalog # USBIN6889-30
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Pore-O-Linked Glycoprotein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: Nuclear Protein Skip
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat