Anti-NFkB, p65, NLS Specific Rabbit Polyclonal Antibody
Catalog # USBIN2303-02A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:NFKB, p65, NLS Specific
- Antigen symbol:NFKB/P65
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Horse,Monkey,Human,Bovine,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Antigen synonyms:p65|NFKB
- Storage buffer:PBS, 0,05% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-3?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: NFkB, p65, NLS Specific
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Horse, Human, Monkey, Mouse