Anti-Neurofibromin Mouse Monoclonal Antibody [clone: 6A137]
USBIN2150-01B
- Antibody type:Primary
- Antigen name:Neurofibromin
- Antigen symbol:NF
- Clonality:Monoclonal
- Clone:6A137
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, containing 0,1% sodium azide
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 0.1-1 ?g/ml; Western Blot: 2-10 ?g/ml
Type: Primary
Antigen: Neurofibromin
Clonality: monoclonal
Clone: 6A137
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat