Order Entry
Ireland
ContactUsLinkComponent
Anti-Melanin Concentrating Hormone Receptor 2 Rabbit Polyclonal Antibody (Rhodamine)
Anti-Melanin Concentrating Hormone Receptor 2 Rabbit Polyclonal Antibody (Rhodamine)
Catalog # USBIM2867-76P
Supplier:  US Biological
Anti-Melanin Concentrating Hormone Receptor 2 Rabbit Polyclonal Antibody (Rhodamine)
Catalog # USBIM2867-76P
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Melanin Concentrating Hormone Receptor 2
  • Antigen symbol:
    MCHR2
  • Clonality:
    Polyclonal
  • Conjugation:
    TRITC (Tetramethylrhodamine Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    liquid
  • Amino acid number:
    237 to 251
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: Melanin Concentrating Hormone Receptor 2
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human