Order Entry
Ireland
ContactUsLinkComponent
Anti-KRT16 Mouse Monoclonal Antibody [clone: LL025]
Anti-KRT16 Mouse Monoclonal Antibody [clone: LL025]
Catalog # USBIK0199-19A
Supplier:  US Biological
Anti-KRT16 Mouse Monoclonal Antibody [clone: LL025]
Catalog # USBIK0199-19A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    keratin 16
  • Antigen symbol:
    KRT16
  • Clonality:
    Monoclonal
  • Clone:
    LL025
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Form:
    liquid
  • Storage buffer:
    10mM PBS, pH 7,4, 0,2% BSA, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulfate precipitation
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: KRT16 (keratin 16)
Clonality: monoclonal
Clone: LL025
Conjugation: unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human