Anti-IL12A Rat Monoclonal Antibody [clone: 11n102]
Catalog # USBII8434-29M
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 12A
- Antigen symbol:IL12A
- Clonality:Monoclonal
- Clone:11n102
- Conjugation:Unconjugated
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:23 to 215
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 8-25 ?g/ml
Type: Primary
Antigen: IL12A (interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35))
Clonality: monoclonal
Clone: 11n102
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse