Anti-Interferon alpha/beta R2 Goat Polyclonal Antibody (APC (Allophycocyanin))
USBII7662-02E
- Antibody type:Primary
- Antigen name:Interferon alpha/beta R2
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Environmentally Preferable:
- Form:liquid
- Storage buffer:saline, 0,5% BSA, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:~0,01 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 Tests
- Pk:100 Tests
Do not freeze!
Type: Primary
Antigen: Interferon alpha/beta R2
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse