Anti-IgG Goat Polyclonal Antibody (TRITC (Rhodamine))
Catalog # USBII1903-89R7
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- Flow cytometry:Yes
- Host:Goat
- Isotype:
- Reactivity:Mouse
- Environmentally Preferable:
- Form:liquid
- Storage buffer:10mMsodium phosphate, 0,15M sodium chloride, pH 7,2, 1% BSA, Protease/IgG free, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse