Anti-HLA Class 2 Antigen DR1 Mouse Monoclonal Antibody [clone: MEM-267]
Catalog # USBIH6099-97R
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:HLA Class 2 Antigen DR1
- Clonality:Monoclonal
- Clone:MEM-267
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 15mM sodium azide, pH 7,4.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: HLA Class 2 Antigen DR1
Clonality: monoclonal
Clone: MEM-267
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human