Anti-HIV-1 p24, Strain IIIB Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5i15]
USBIH6003-33D3
- Antibody type:Primary
- Antigen symbol:HIV-1 p24, Strain IIIB
- Clonality:Monoclonal
- Clone:5i15
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:10mM Tris, pH 8.0. No s or
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Dot Blot
- Purification:purified by gel exclusion chromatography
- Size:50 Tests
- Pk:50 Tests
Type: Primary
Antigen: HIV-1 p24, Strain IIIB
Clonality: monoclonal
Clone: 5i15
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: