Anti-GMR-alpha Mouse monoclonal antibody [clone: 4H1]
Catalog # USBIG8951-31G
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:GMR-alpha
- Clonality:Monoclonal
- Clone:4H1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: GMR-alpha
Clonality: monoclonal
Clone: 4H1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human