Anti-G Protein Coupled Receptor 54 Rabbit polyclonal antibody Rhodamine
Catalog # USBIG8600-58G3
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:G Protein Coupled Receptor 54
- Antigen symbol:GPR54
- Clonality:Polyclonal
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Environmentally Preferable:
- Form:liquid
- Amino acid number:375 to 396
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: G Protein Coupled Receptor 54
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse