Anti-FURIN Rabbit Polyclonal Antibody
Catalog # USBIF9200-01A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Furin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Horse,Human,Bovine,Dog,Rat,Mouse
- Environmentally Preferable:
- Form:liquid
- Amino acid number:740 to 790
- Storage buffer:PBS, 0,05% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: FURIN (furin (paired basic amino acid cleaving enzyme))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Horse, Human, Mouse, Rat