Anti-Folate Binding Protein Goat Polyclonal Antibody (Biotin)
USBIF5750-10A
- Antibody type:Primary
- Antigen name:Folate Binding Protein
- Antigen symbol:FBP
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Bovine
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 10mg/ml BSA, 0,01% sodium azide, 40% glycerol.
- Storage temperature:Cool
- Concentration:~10 mg/ml (UV280)
- Shipping temperature:Ice
- Purification:Purified from monospecific antiserum by delipidation, fractionation and ion exchange chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:5000-1:25000
Type: Primary
Antigen: Folate Binding Protein
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine