Anti-Epithelial Specific Antigen Mouse Monoclonal Antibody [clone: 5G99]
Catalog # USBIE3414-11K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Epithelial Specific Antigen
- Antigen symbol:EpCAM
- Clonality:Monoclonal
- Clone:5G99
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Format:Cell Culture supernatant
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Type: Primary
Antigen: Epithelial Specific Antigen
Clonality: monoclonal
Clone: 5G99
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human