Anti-CTP Phosphocholine cytidylyltransferase Rabbit polyclonal antibody
Catalog # USBIC7948-35
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CTP Phosphocholine cytidylyltransferase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:10mMsodium HEPES pH 7,5, 150mM NaCl, 100ug/ml BSA and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: CTP Phosphocholine cytidylyltransferase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human