Anti-CRACC Rat Monoclonal Antibody [clone: 9H102]
Catalog # USBIC7906-72C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CRACC
- Clonality:Monoclonal
- Clone:9H102
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:22 to 224
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 25 ?g/ml
Type: Primary
Antigen: CRACC
Clonality: Monoclonal
Clone: 9H102
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse