Order Entry
Ireland
ContactUsLinkComponent
Anti-Contactin-associated protein 1 Mouse Monoclonal Antibody [clone: 11C235]
Anti-Contactin-associated protein 1 Mouse Monoclonal Antibody [clone: 11C235]
Catalog # USBIC7865-19A
Supplier:  US Biological
Anti-Contactin-associated protein 1 Mouse Monoclonal Antibody [clone: 11C235]
Catalog # USBIC7865-19A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Contactin-associated Protein 1
  • Clonality:
    Monoclonal
  • Clone:
    11C235
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    0,1M Tris-glycine, pH 7,4, 150mM sodium chloride, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 1:300; Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: Contactin-associated protein 1
Clonality: Monoclonal
Clone: 11C235
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Rat