Anti-CIT Rabbit polyclonal antibody
USBIC5804-20
- Antibody type:Primary
- Antigen name:citron (rho-interacting, serine/threonine kinase 21)
- Antigen symbol:CIT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Epitope:internal
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: CIT (citron (rho-interacting, serine/threonine kinase 21))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human