Anti-CD2 Mouse Monoclonal Antibody [clone: BL-TP2/1]
Catalog # USBIC2253-02J
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CD2 molecule
- Antigen symbol:CD2
- Clonality:Monoclonal
- Clone:BL-TP2/1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Format:ascites
- Form:lyophilized powder
- Storage buffer:sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: CD2 (CD2 molecule)
Clonality: Monoclonal
Clone: BL-TP2/1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human