Anti-CASQ2 Mouse Monoclonal Antibody [clone: 1C6]
Catalog # USBIC1036-07A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:calsequestrin 2 (cardiac muscle)
- Antigen symbol:CASQ2
- Clonality:Monoclonal
- Clone:1C6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:20 to 400
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: CASQ2 (calsequestrin 2 (cardiac muscle))
Clonality: Monoclonal
Clone: 1C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human