Anti-Bacillus cereus/Bacillus subtilis, Spores Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIB0003-27B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Bacillus cereus/Bacillus subtilis, Spores
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~4-5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:800
Type: Primary
Antigen: Bacillus cereus/Bacillus subtilis, Spores
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: