Anti-AP2S1 Rabbit Monoclonal Antibody [clone: 9H292]
USBIA2298-51T
- Antibody type:Primary
- Antigen name:adaptor-related protein complex 2, sigma 1 subunit
- Antigen symbol:AP2S1
- Clonality:Monoclonal
- Clone:9H292
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:Supernatant
- Form:liquid
- Storage buffer:PBS, 0,05% BSA, 0,01% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Recommended dilution: Flow Cytometry: 1:200; Western Blot: 1:1000-1:5000; Immunoprecipitation: 1:10
Type: Primary
Antigen: AP2S1 (adaptor-related protein complex 2, sigma 1 subunit)
Clonality: monoclonal
Clone: 9H292
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat