Anti-AIB 1 Mouse Monoclonal Antibody [clone: 6D102]
Catalog # USBIA0857-06A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Nuclear receptor coactivator 3
- Antigen symbol:NCOA3
- Clonality:Monoclonal
- Clone:6D102
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Antigen synonyms:NCOA3
- Amino acid number:605 to 1294
- Storage buffer:PBS containing BSA and 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: AIB 1
Clonality: monoclonal
Clone: 6D102
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human