Anti-NOGO-A Rabbit Polyclonal Antibody
Catalog # USBI144977
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Nogo A
- Antigen symbol:NOGO-A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:5mgBSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mgthimerosal, 0,05mg NaN3.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: NOGO-A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat