Anti-OLFM4 Rabbit Polyclonal Antibody
50 µg
Catalog # USBI136303
Supplier: US Biological
Jump to:
Product Details & Documents
Recommended dilution: Immunochemistry: 15-20 ?g/ml
Type: Primary
Antigen: OLFM4 (olfactomedin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
Specifications
- Size:50 µg
- Clonality:Polyclonal
- Purification:purified by immunoaffinity chromatography
- Antibody type:Primary
- Form:liquid
- Storage temperature:Cool
- Host:Rabbit
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Antigen name:olfactomedin 4
- Cat. No.:USBI136303
- Reactivity:Human
- Conjugation:Unconjugated
- Antigen symbol:OLFM4
- Storage buffer:PBS, 0,1% sodium azide.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



