Anti-Dopamine Beta Hydroxylase Sheep Polyclonal Antibody
Catalog # USBI136040
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Dopamine Beta Hydroxylase
- Antigen symbol:DBH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Form:liquid
- Storage buffer:HEPES (pH 7,5), 150mM NaCl, 100ug per ml BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:100
Type: Primary
Antigen: Dopamine Beta Hydroxylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Sheep
Isotype:
Reactivity: Human