Anti-UTRN Mouse Monoclonal Antibody [clone: 5G6]
Catalog # USBI135189
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:utrophin
- Antigen symbol:UTRN
- Clonality:Monoclonal
- Clone:5G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:3328 to 3433
- Storage buffer:PBS, pH 7,4.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: UTRN (utrophin)
Clonality: monoclonal
Clone: 5G6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human