Anti-RHOQ Mouse Monoclonal Antibody [clone: 2C1]
Catalog # USBI132565
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Ras Homolog Family Member Q
- Antigen symbol:RHOQ
- Clonality:Monoclonal
- Clone:2C1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Amino acid number:106 to 205
- Storage buffer:PBS, pH 7,2.
- Sequence:ELKEYAPNVPFLLIGTQIDLRDDPKTLARLNDMKEKPICVEQGQKLAKEIGACCYVECSALTQKGLKTVFDEAIIAILTPKKHTVKKRIGSRCINCCLI*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: RHOQ (ras homolog family member Q)
Clonality: monoclonal
Clone: 2C1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human