Anti-PTPN4 Mouse Monoclonal Antibody [clone: 3C8]
USBI132067
- Antibody type:Primary
- Antigen name:protein tyrosine phosphatase, non-receptor type 4 (megakaryocyte)
- Antigen symbol:PTPN4
- Clonality:Monoclonal
- Clone:3C8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:361 to 461
- Storage buffer:PBS, pH 7,2.
- Sequence:KPLARKLMDWEVVSRNSISDDRLETQSLPSRSPPGTPNHRNSTFTQEGTRLRPSSVGHLVDHMVHTSPSEVFVNQRSPSSTQANSIVLESSPSQETPGDG
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: PTPN4 (protein tyrosine phosphatase, non-receptor type 4 (megakaryocyte))
Clonality: monoclonal
Clone: 3C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human