Anti-PKD1L2 Mouse Monoclonal Antibody [clone: 3F6]
USBI131387
- Antibody type:Primary
- Antigen name:Polycystic Kidney Disease 1-like 2
- Antigen symbol:PKD1L2
- Clonality:Monoclonal
- Clone:3F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites
- Form:liquid
- Storage buffer:ascites fluid.
- Sequence:MGEDSPVAMFSWYLDNTPTEQAEPLPDACRLRGFWPRSLTLLQSNTSTLLLNSSFLQSRGEVIRIRATALTRHAYGEDTYVISTVPPREVPACTIAPEEGTVLTSFAIFCNASTALGPLEFCFCLESGSCLHCGPEPALPSVYLPLGEENNDFVLTVVISATNRAGDTQQTQAMAKVALGDTCVEDVAFQAAVSEKIPTALQGEGGPEQLLQLAKAVSSMLNQEHESQGSGQSLSIDVRQKVREHVLGSLSAVTTGLEDVQRVQELAEVLREVTCRSKELTPSAQGSCMGDSWEGAPPAAHVSHAR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: PKD1L2 (polycystic kidney disease 1-like 2)
Clonality: monoclonal
Clone: 3F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human