Anti-PDCD11 Mouse Monoclonal Antibody [clone: 3E12]
Catalog # USBI131030
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Programmed cell death 11
- Antigen symbol:PDCD11
- Clonality:Monoclonal
- Clone:3E12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MANLEESFPRGGTRKIHKPEKAFQQSVEQDNLFDISTEEGSTKRKKSQKGPAKTKKLKIEKRESSKSAREKFEILSVESLCEGMRILGCVKEVNELELVISLPNGLQGFV
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PDCD11 (programmed cell death 11)
Clonality: monoclonal
Clone: 3000000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human