Anti-KLK4 Mouse Monoclonal Antibody [clone: 2A4]
Catalog # USBI128713
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:kallikrein-related peptidase 4
- Antigen symbol:KLK4
- Clonality:Monoclonal
- Clone:2A4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:159 to 254
- Storage buffer:PBS, pH 7,2.
- Sequence:GRMPTVLQCVNVSVVSEEVCSKLYDPLYHPSMFCAGGGQDQKDSCNGDSGGPLICNGYLQGLVSFGKAPCGQVGVPGVYTNLCKFTEWIEKTVQAS
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: KLK4 (kallikrein-related peptidase 4)
Clonality: monoclonal
Clone: 2A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human