Anti-Hexokinase, Type IV Mouse Monoclonal Antibody [clone: 3C7]
Catalog # USBI127826
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Hexokinase 4
- Antigen symbol:HK4
- Clonality:Monoclonal
- Clone:3C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:GLRPSTTDCDIVRRACESVSTRAAHMCSAGLAGVINRMRESRSEDVMRITVGVDGSVYKLHPSFKERFHASVRRLTPSCEITFIESEEGSGRGAALVSAVACKKACMLGQ
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Hexokinase, Type IV
Clonality: monoclonal
Clone: 3C7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human