Order Entry
Ireland
ContactUsLinkComponent
Anti-GMPR Mouse Monoclonal Antibody [clone: 3G12]
Anti-GMPR Mouse Monoclonal Antibody [clone: 3G12]
Catalog # USBI127403
Supplier:  US Biological
Anti-GMPR Mouse Monoclonal Antibody [clone: 3G12]
Catalog # USBI127403
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Guanosine monophosphate reductase
  • Antigen symbol:
    GMPR
  • Clonality:
    Monoclonal
  • Clone:
    3G12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgM kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Format:
    ascites
  • Form:
    liquid
  • Amino acid number:
    1 to 120
  • Storage buffer:
    ascites fluid.
  • Sequence:
    MPRIDADLKLDFKDVLLRPKRSSLKSRAEVDLERTFTFRNSKQTYSGIPIIVANMDTVGTFEMAAVMSQHSMFTAIHKHYSLDDWKLFATNHPECLQNVAVSSGSGQNDLEKMTSILEAV
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: GMPR (guanosine monophosphate reductase)
Clonality: monoclonal
Clone: 3G12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human