Anti-DDOST Mouse Monoclonal Antibody [clone: 2D7]
USBI125716
- Antibody type:Primary
- Antigen name:dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit (non-catalytic)
- Antigen symbol:DDOST
- Clonality:Monoclonal
- Clone:2D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:328 to 427
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: DDOST (dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit (non-catalytic))
Clonality: monoclonal
Clone: 2D7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human