Anti-CPA3 Mouse Monoclonal Antibody [clone: 2D1]
Catalog # USBI125270
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:carboxypeptidase A3 (mast cell)
- Antigen symbol:CPA3
- Clonality:Monoclonal
- Clone:2D1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Amino acid number:318 to 417
- Storage buffer:PBS, pH 7,2.
- Sequence:SKLPPNHEDLAKVAKIGTDVLSTRYETRYIYGPIESTIYPISGSSLDWAYDLGIKHTFAFELRDKGKFGFLLPESRIKPTCRETMLAVKFIAKYILKHTS
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CPA3 (carboxypeptidase A3 (mast cell))
Clonality: monoclonal
Clone: 2D1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human