Anti-ADAMDEC1 Mouse Monoclonal Antibody [clone: 6C4]
Catalog # USBI122977
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ADAM-like, decysin 1
- Antigen symbol:ADAMDEC1
- Clonality:Monoclonal
- Clone:6C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:361 to 471
- Storage buffer:PBS, pH 7,2.
- Sequence:PDVPFNTKCPSGSCVMNQYLSSKFPKDFSTSCRAHFERYLLSQKPKCLLQAPIPTNIMTTPVCGNHLLEVGEDCDCGSPKECTNLCCEALTCKLKPGTDCGGDAPNHTTE*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 3?g/ml
Type: Primary
Antigen: ADAMDEC1 (ADAM-like, decysin 1)
Clonality: monoclonal
Clone: 6C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human