Order Entry
Ireland
ContactUsLinkComponent
Anti-TC-PTP Mouse Monoclonal Antibody
Anti-TC-PTP Mouse Monoclonal Antibody
Catalog # MEDMMM-0018-P
Supplier:  MédiMabs
Anti-TC-PTP Mouse Monoclonal Antibody
Catalog # MEDMMM-0018-P
Supplier:  MédiMabs

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TC-PTP
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: TC-PTP
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Mouse