Anti-FGF-21 Mouse Monoclonal Antibody [clone: FG348-1]
Catalog # USBI044799
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:FGF-21
- Clonality:Monoclonal
- Clone:FG348-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:0,2um-filtered PBS, pH 7,4
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: FGF-21
Clonality: Monoclonal
Clone: FG348-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: