Anti-PDK3 Mouse Monoclonal Antibody [clone: 431CT6.1]
Catalog # USBI031186
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:pyruvate dehydrogenase kinase, isozyme 3
- Antigen symbol:PDK3
- Clonality:Monoclonal
- Clone:431CT6.1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-800
Type: Primary
Antigen: PDK3 (pyruvate dehydrogenase kinase, isozyme 3)
Clonality: Monoclonal
Clone: 431CT6.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human