Anti-MOUSE Serum Albumin Rabbit Polyclonal Antibody
ROCK600-401-254
- Antibody type:Primary
- Antigen name:albumin
- Antigen symbol:ALB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Gene ID:11657
- Antigen synonyms:Alb1|Alb-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: MOUSE Serum Albumin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse