Anti-Serotonin (5-HT) Transporter (602-622) Rabbit Polyclonal Antibody
Catalog # PC177L-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:5-HT Transporter
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:Serotonin
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Serotonin (5-HT) Transporter (602-622)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: